3-Pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

P1673-5G P1673-1G P1673-25G
TCP1673-5GEA 82.16 CAD
TCP1673-5G TCP1673-1G TCP1673-25G
3-Pyridineboronic acid (contains varying amounts of Anhydride)
3-Pyridineboronic acid

CAS Number: 1692-25-7
MDL Number: MFCD00674177
Molecular Formula: C5H6BNO2
Molecular Weight: 122.92
Form: Crystal
Color: White


Other Product Names / Synonyms: Pyridine-3-boronic Acid

Formula: C₅H₆BNO₂
MDL Number: MFCD00674177
CAS Number: 1692-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72112dab2471ed995e1b463122a655f959f9785f&vid=72112dab2471ed995e1b463122a655f959f9785f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR