(S)-(-)-Hydroxypyrrolidine ≥97.0%

Supplier: TCI America

Synonyms: (S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine

P1667-5G P1667-1G P1667-25G
TCP1667-5GEA 221.52 CAD
TCP1667-5G TCP1667-1G TCP1667-25G
(S)-(-)-Hydroxypyrrolidine ≥97.0%
(S)-(-)-Hydroxypyrrolidine

CAS Number: 100243-39-8
MDL Number: MFCD00192426
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 110
Flash Point (°C): 109
Specific Gravity (20/20): 1.08
Specific rotation [a]20/D: -5.7 deg (C=3, MeOH)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-3-Hydroxypyrrolidine

Formula: C₄H₉NO
Boiling Pt: 110 °C
Density: 1.08 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00192426
CAS Number: 100243-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7487dc1725379310cde442ca1f7a0d66ff880712&vid=7487dc1725379310cde442ca1f7a0d66ff880712&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR