1,4'-Bipiperidine ≥98.0%
Supplier: TCI America
P1638-5G
P1638-1G
P1638-25G
TCP1638-5GEA
194.34
CAD
TCP1638-5G
TCP1638-1G
TCP1638-25G
1,4'-Bipiperidine ≥98.0%
1,4'-Bipiperidine
CAS Number: 4897-50-1
Molecular Formula: C10H20N2
Molecular Weight: 168.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 66
Formula:
C₁₀H₂₀N₂ Melting Pt: 66 °C |
MDL Number:
MFCD00006475 CAS Number: 4897-50-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...