(R)-(+)-Hydroxypyrrolidine ≥98.0%

Supplier: TCI America

Synonyms: (R)-Pyrrolidin-3-ol, (R)-(+)-3-Pyrrolidinol, (R)-(+)-3-Hydroxypyrrolidine

P1608-1G P1608-5G
TCP1608-1GEA 82.63 CAD
TCP1608-1G TCP1608-5G
(R)-(+)-Hydroxypyrrolidine ≥98.0%
(R)-(+)-Hydroxypyrrolidine

CAS Number: 2799-21-5
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 70
Flash Point (°C): 113
Freezing point (°C): 30
Specific rotation [a]20/D: 6 deg (C=3, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-3-Hydroxypyrrolidine

Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 70 °C
Flash Pt: 113 °C
MDL Number: MFCD00145220
CAS Number: 2799-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a1552c0f4aea5bebbe1da4dcbcf315cc0a6dcc0&vid=0a1552c0f4aea5bebbe1da4dcbcf315cc0a6dcc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR