Potassium (4-methylphenyl)trifluoroborate ≥98.0% (by HPLC)

Supplier: TCI America

P1583-1G P1583-5G
TCP1583-1GEA 96.38 CAD
TCP1583-1G TCP1583-5G
Potassium (4-methylphenyl)trifluoroborate ≥98.0% (by HPLC)
Potassium (4-methylphenyl)trifluoroborate

CAS Number: 216434-82-1
MDL Number: MFCD01318171
Molecular Formula: C7H7BF3K
Molecular Weight: 198.04
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 294

Formula: C₇H₇BF₃K
Melting Pt: 294 °C
MDL Number: MFCD01318171
CAS Number: 216434-82-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b17bbcf878ed1b5772dafe37acade01e7bb3a8f8&vid=b17bbcf878ed1b5772dafe37acade01e7bb3a8f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR