Potassium phenyltrifluoroborate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Potassium trifluoro(phenyl)borate, Phenyltrifluoroboric acid potassium salt

P1582-5G P1582-1G P1582-25G
TCP1582-5GEA 270.18 CAD
TCP1582-5G TCP1582-1G TCP1582-25G
Potassium phenyltrifluoroborate ≥98.0% (by HPLC, titration analysis)
Potassium phenyltrifluoroborate

CAS Number: 153766-81-5
MDL Number: MFCD01318172
Molecular Formula: C6H5BF3K
Molecular Weight: 184.01
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 301

Formula: C₆H₅BF₃K
MW: 184.01 g/mol
Melting Pt: 301 °C
MDL Number: MFCD01318172
CAS Number: 153766-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48f8cfbe605cb85dfdf6e0e3b71b9a7483f31064&vid=48f8cfbe605cb85dfdf6e0e3b71b9a7483f31064&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR