1-Phenylcyclopentanecarboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 1-Phenyl-1-cyclopentanecarboxylic acid

P1487-25G P1487-5G
TCP1487-25GEA 121.66 CAD
TCP1487-25G TCP1487-5G
1-Phenylcyclopentanecarboxylic acid ≥98.0%
1-Phenylcyclopentanecarboxylic acid

CAS Number: 77-55-4
MDL Number: MFCD00001372
Molecular Formula: C12H14O2
Molecular Weight: 190.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 159

Formula: C₁₂H₁₄O₂
Melting Pt: 159 °C
MDL Number: MFCD00001372
CAS Number: 77-55-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR