Potassium vinyltrifluoroborate ≥97.0% (by gravimetric analysis)

Supplier: TCI America

Synonyms: Potassium trifluoro(vinyl)borate

P1479-5G P1479-1G P1479-25G
TCP1479-5GEA 69.52 CAD
TCP1479-5G TCP1479-1G TCP1479-25G
Potassium vinyltrifluoroborate ≥97.0% (by gravimetric analysis)
Potassium vinyltrifluoroborate

CAS Number: 13682-77-4
Molecular Formula: C2H4BF3
Molecular Weight: 133.95
Purity/Analysis Method: >97.0% (W)
Form: Crystal
Color: White

Formula: C₂H₃BF₃K
MW: 133.95 g/mol
MDL Number: MFCD02093335
CAS Number: 13682-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73787380c4bc7729d562386a28a432121a5476b2&vid=73787380c4bc7729d562386a28a432121a5476b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (W)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR