Potassium tris(1-pyrazolyl)borohydride ≥97.0% (by titrimetric analysis)
Supplier: TCI America
P1440-5G
P1440-1G
TCP1440-5GEA
245.83
CAD
TCP1440-5G
TCP1440-1G
Potassium tris(1-pyrazolyl)borohydride ≥97.0% (by titrimetric analysis)
Potassium tris(1-pyrazolyl)borohydride
CAS Number: 18583-60-3
MDL Number: MFCD00040346
Molecular Formula: C9H10BKN6
Molecular Weight: 252.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Potassium Hydrotris(1-pyrazolyl)borate
Formula:
C₉H₁₀BKN₆ |
MDL Number:
MFCD00040346 CAS Number: 18583-60-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...