Propargyltriphenylphosphonium bromide ≥98.0%
Supplier: TCI America
Synonyms:
2-Propinyl-triphenylphosphonium bromide, Triphenyl(prop-2-yn-1-yl)phosphonium bromide
CAS Number: 2091-46-5
MDL Number: MFCD00011811
Molecular Formula: C21H18BrP
Molecular Weight: 381.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Propargyltriphenylphosphonium Bromide, Triphenyl(2-propynyl)phosphonium Bromide
Formula:
C₂₁H₁₈BrP MW: 381.26 g/mol |
MDL Number:
MFCD00011811 CAS Number: 2091-46-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...