1,4-Phenylenediacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P1433-5G P1433-25G
TCP1433-5GEA 121.66 CAD
TCP1433-5G TCP1433-25G
1,4-Phenylenediacetic acid ≥98.0% (by HPLC, titration analysis)
1,4-Phenylenediacetic acid

CAS Number: 7325-46-4
MDL Number: MFCD00004354
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00004354
CAS Number: 7325-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eec8a4e86fcea289bcdcea141ceba1be89643431&vid=eec8a4e86fcea289bcdcea141ceba1be89643431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR