Poly(styrene) on resin, cross-linked with 1% DVB (100-200mesh)

Supplier: TCI America

P1432-25G P1432-100G
TCP1432-25GEA 191.18 CAD
TCP1432-25G TCP1432-100G
Poly(styrene) on resin, cross-linked with 1% DVB (100-200mesh)
Poly(styrene)

MDL Number: MFCD00146442
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Resins (Combinatorial Chemistry)

Formula: C₈H₈
MDL Number: MFCD00084450
CAS Number: 9003-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e674c16fc30d1c9c7e61788abdf1c1b69e658f8e&vid=786033e5917bb80f33137486a9d99aea0c6d2182&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR