Ethyl pentafluorobenzoate ≥98.0%
Supplier: TCI America
P1419-5G
P1419-25G
TCP1419-5GEA
42.66
CAD
TCP1419-5G
TCP1419-25G
Ethyl pentafluorobenzoate ≥98.0%
Ethyl pentafluorobenzoate
CAS Number: 4522-93-4
MDL Number: MFCD00039211
Molecular Formula: C9H5F5O2
Molecular Weight: 240.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 113
Flash Point (°C): 81
Specific Gravity (20/20): 1.43
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Ethyl Perfluorobenzoate, Pentafluorobenzoic Acid Ethyl Ester, Perfluorobenzoic Acid Ethyl Ester
Formula:
C₉H₅F₅O₂ Boiling Pt: 113 °C Density: 1.43 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00039211 CAS Number: 4522-93-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...