Tri-o-tolyl phosphite ≥97.0%

Supplier: TCI America

P1416-1G P1416-5G
TCP1416-1GEA 112.18 CAD
TCP1416-1G TCP1416-5G
Tri-o-tolyl phosphite ≥97.0%
Tri-o-tolyl phosphite

CAS Number: 2622-08-4
MDL Number: MFCD00014911
Molecular Formula: C21H21O3P
Molecular Weight: 352.37
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 238
Specific Gravity (20/20): 1.14
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Phosphorous Acid Tri-o-cresyl Ester, Phosphorous Acid Tris(2-methylphenyl) Ester, Phosphorous Acid Tri-o-tolyl Ester, Tri-o-cresyl Phosphite, Tris(2-methylphenyl) Phosphite

Formula: C₂₁H₂₁O₃P
Boiling Pt: 238 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00014911
CAS Number: 2622-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8f668d6bead0f4895d0a1fa6f63da808a5053e7&vid=b8f668d6bead0f4895d0a1fa6f63da808a5053e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR