2-Phenylisobutyric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1409-250G P1409-25G
TCP1409-250GEA 842.14 CAD
TCP1409-250G TCP1409-25G
2-Phenylisobutyric acid ≥98.0% (by GC, titration analysis)
2-Phenylisobutyric acid

CAS Number: 826-55-1
MDL Number: MFCD00014332
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 79


Other Product Names / Synonyms: 2-Methyl-2-phenylpropionic Acid

Formula: C₁₀H₁₂O₂
Melting Pt: 79 °C
MDL Number: MFCD00014332
CAS Number: 826-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26de338dd5a08ff11061fd841248b6e8ae7644b7&vid=26de338dd5a08ff11061fd841248b6e8ae7644b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR