L-Pipecolic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-(-)-2-Piperidinecarboxylic Acid, H-HomoPro-OH, (S)-Piperidine-2-carboxylic acid, L-Pipecolinic Acid, L-Homoproline

P1404-1G P1404-5G
TCP1404-1GEA 148.84 CAD
TCP1404-1G TCP1404-5G
L-Pipecolic acid ≥97.0% (by titrimetric analysis)
L-Pipecolic acid

CAS Number: 3105-95-1
MDL Number: MFCD00005981
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -27 deg (C=4, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Chiral Building Blocks, Bioscience, Other Plant Hormones, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: L-Homoproline, L-Pipecolinic Acid, (S)-(-)-2-Piperidinecarboxylic Acid

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD00005981
CAS Number: 3105-95-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9859abe73953a1f3eb30ea75de802f07564ee160&vid=9859abe73953a1f3eb30ea75de802f07564ee160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR