(4-Pyridyl)acetic acid ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Ethyl (4-pyridyl)acetate, Ethyl 4-pyridylacetate, Ethyl 4-pyridineacetate

P1399-25G P1399-5G
TCP1399-25GEA 443.03 CAD
TCP1399-25G TCP1399-5G
(4-Pyridyl)acetic acid ethyl ester ≥98.0%
(4-Pyridyl)acetic acid ethyl ester

CAS Number: 54401-85-3
MDL Number: MFCD00051830
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 128
Flash Point (°C): 113
Freezing point (°C): 19
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: Ethyl 4-Pyridineacetate, 4-Pyridylacetic Acid Ethyl Ester, 4-Pyridineacetic Acid Ethyl Ester

Formula: C₉H₁₁NO₂
MW: 165.19
Boiling Pt: 128 °C
Density: 1.09 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00051830
CAS Number: 54401-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=680a6ee300f6d06d5d122d9183c0e72ff2763b5b&vid=680a6ee300f6d06d5d122d9183c0e72ff2763b5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR