(4-Chlorobutyl)benzene ≥98.0%
Supplier: TCI America
P1389-25G
P1389-5G
TCP1389-25GEA
538.78
CAD
TCP1389-25G
TCP1389-5G
(4-Chlorobutyl)benzene ≥98.0%
(4-Chlorobutyl)benzene
CAS Number: 4830-93-7
MDL Number: MFCD00037117
Molecular Formula: C10H13Cl
Molecular Weight: 168.66
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 104
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: (4-Chlorobutyl)benzene, 1-Chloro-4-phenylbutane
Formula:
(C₆H₅)(CH₂)₄Cl MW: 168.67 g/mol Density: 1.03 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00037117 CAS Number: 4830-93-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...