1-Bromo-4-phenylbutane ≥97.0%

Supplier: TCI America

P1388-5G P1388-25G
TCP1388-5GEA 95.04 CAD
TCP1388-5G TCP1388-25G
1-Bromo-4-phenylbutane ≥97.0%
1-Bromo-4-phenylbutane

CAS Number: 13633-25-5
MDL Number: MFCD00154988
Molecular Formula: C10H13Br
Molecular Weight: 213.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.27


Other Product Names / Synonyms: (4-Bromobutyl)benzene, 1-Bromo-4-phenylbutane

Formula: C₁₀H₁₃Br
Density: 1.27 (20 °C)
MDL Number: MFCD00154988
CAS Number: 13633-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c09565b585b882db7e351a9538315c41ba8955b7&vid=c09565b585b882db7e351a9538315c41ba8955b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR