L-Prolinamide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

P1382-25G P1382-5G P1382-1G
TCP1382-25GEA 192.76 CAD
TCP1382-25G TCP1382-5G TCP1382-1G
L-Prolinamide ≥97.0% (by titrimetric analysis)
L-Prolinamide

CAS Number: 7531-52-4
MDL Number: MFCD00005253
Molecular Formula: C5H10N2O
Molecular Weight: 114.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 99
Specific rotation [a]20/D: -106 deg (C=2, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acid Amides
Other Product Names / Synonyms: (S)-2-Pyrrolidinecarboxamide

Formula: C₅H₁₀N₂O
Melting Pt: 99 °C
MDL Number: MFCD00005253
CAS Number: 7531-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24ecccf359e003aac337c4b02c564a05461a05d3&vid=24ecccf359e003aac337c4b02c564a05461a05d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR