(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol ≥98.0%

Supplier: TCI America

Synonyms: (1S,2R)-N,N-Tetramethylenenorephedrine

P1375-1G P1375-5G
TCP1375-001GEA 80.58 CAD
TCP1375-001G TCP1375-5G
(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol ≥98.0%
(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol

CAS Number: 123620-80-4
MDL Number: MFCD02093492
Molecular Formula: C13H19NO
Molecular Weight: 205.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 48
Specific rotation [a]20/D: -14.5 deg (C=2, CHCl3)


Other Product Names / Synonyms: (1S,2R)-N,N-Tetramethylenenorephedrine

Formula: C₁₃H₁₉NO
MW: 205.30 g/mol
Melting Pt: 48 °C
MDL Number: MFCD02093492
CAS Number: 123620-80-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0f37a30de5b93d7bc31608da97bfa897d28623f&vid=b0f37a30de5b93d7bc31608da97bfa897d28623f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR