Methyl Phenylsulfonylacetate ≥95.0%

Supplier: TCI America

Synonyms: Phenylsulphonylacetic Acid Methyl Ester, Phenylsulfonylacetic Acid Methyl Ester

P1353-5G P1353-25G
TCP1353-005GEA 71.1 CAD
TCP1353-005G TCP1353-25G
Methyl Phenylsulfonylacetate ≥95.0%
Methyl Phenylsulfonylacetate

CAS Number: 34097-60-4
MDL Number: MFCD00007555
Molecular Formula: C9H10O4S
Molecular Weight: 214.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Flash Point (°C): 78
Specific Gravity (20/20): 1.30


Other Product Names / Synonyms: Phenylsulfonylacetic Acid Methyl Ester

Formula: C₉H₁₀O₄S
MW: 214.24 g/mol
Density: 1.30 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00007555
CAS Number: 34097-60-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9671eb9c980a5c9596a34043817a8b24fbab18e5&vid=9671eb9c980a5c9596a34043817a8b24fbab18e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR