4-Phenyl-3-butyn-2-one ≥97.0%
Supplier: TCI America
P1336-5G
P1336-1G
TCP1336-005GEA
421.86
CAD
TCP1336-005G
TCP1336-001G
4-Phenyl-3-butyn-2-one ≥97.0%
4-Phenyl-3-butyn-2-one
CAS Number: 1817-57-8
MDL Number: MFCD00008776
Molecular Formula: C10H8O
Molecular Weight: 144.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 130
Flash Point (°C): 95
Specific Gravity (20/20): 1.02
Formula:
C₁₀H₈O Boiling Pt: 130 °C Density: 1.02 (20 °C) Flash Pt: 95 °C |
MDL Number:
MFCD00008776 CAS Number: 1817-57-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...