Phenyl-β-D-galactopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
P1326-1G
P1326-5G
TCP1326-001GEA
117.39
CAD
TCP1326-001G
TCP1326-005G
Phenyl-β-D-galactopyranoside ≥98.0% (by HPLC)
Phenyl-beta-D-galactopyranoside
CAS Number: 2818-58-8
MDL Number: MFCD00063258
Molecular Formula: C12H16O6
Molecular Weight: 256.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 148
Specific rotation [a]20/D: -42 deg (C=2.3, H2O)
Formula:
C₁₂H₁₆O₆ Melting Pt: 148 °C |
MDL Number:
MFCD00063258 CAS Number: 2818-58-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...