γ-Phenyl-γ-butyrolactone ≥97.0%

Supplier: TCI America

P1321-5G P1321-25G
TCP1321-005GEA 180.12 CAD
TCP1321-005G TCP1321-25G
γ-Phenyl-γ-butyrolactone ≥97.0%
gamma-Phenyl-gamma-butyrolactone

CAS Number: 1008-76-0
MDL Number: MFCD00005398
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 306
Melting point (°C): 36
Flash Point (°C): 113

Formula: C₁₀H₁₀O₂
Boiling Pt: 306 °C
Melting Pt: 36 °C
Flash Pt: 113 °C
MDL Number: MFCD00005398
CAS Number: 1008-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0280ee0045f2b199a9677042de82014e0beca31&vid=b0280ee0045f2b199a9677042de82014e0beca31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR