2-(Pyridin-4-yl)acetic acid hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 4-Pyridylacetic acid hydrochloride

P1317-25G P1317-5G
TCP1317-25GEA 97.49 CAD
TCP1317-25G TCP1317-005G
2-(Pyridin-4-yl)acetic acid hydrochloride ≥98.0%
2-(Pyridin-4-yl)acetic acid hydrochloride

CAS Number: 6622-91-9
MDL Number: MFCD00012827
Molecular Formula: C7H7NO2
Molecular Weight: 173.60
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-Pyridineacetic Acid Hydrochloride

Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
MDL Number: MFCD00012827
CAS Number: 6622-91-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92d1e3d5b40e67bff899303b34248202d0310528&vid=92d1e3d5b40e67bff899303b34248202d0310528&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR