Trimethyl phosphonoacetate ≥96.0%

Supplier: TCI America

Synonyms: Methyl 2-(dimethoxyphosphoryl)acetate, Dimethyl methoxycarbonylmethanephosphonate

P1265-250G P1265-25G
TCP1265-250GEA 381.14 CAD
TCP1265-250G TCP1265-025G
Trimethyl phosphonoacetate ≥96.0%
Trimethyl phosphonoacetate

CAS Number: 5927-18-4
MDL Number: MFCD00008452
Molecular Formula: C5H11O5P
Molecular Weight: 182.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 108
Flash Point (°C): 70
Specific Gravity (20/20): 1.27


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Dimethylphosphonoacetic Acid Methyl Ester, Methyl Dimethylphosphonoacetate, Phosphonoacetic Acid Trimethyl Ester

Formula: C₅H₁₁O₅P
Boiling Pt: 108 °C
Density: 1.27 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00008452
CAS Number: 5927-18-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8d53f6bb2a6db730ebcb6beaa430dafc205078d&vid=e8d53f6bb2a6db730ebcb6beaa430dafc205078d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR