N,N-Bis(trifluoromethanesulfonyl)aniline ≥98.0%

Supplier: TCI America

Synonyms: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulfonimide), N-Phenylbis(trifluoromethanesulphonimide), 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulfonyl)methanesulfonamide

P1257-25G P1257-5G
TCP1257-025GEA 132.25 CAD
TCP1257-025G TCP1257-005G
N,N-Bis(trifluoromethanesulfonyl)aniline ≥98.0%
N,N-Bis(trifluoromethanesulphonyl)aniline

[Triflating Reagent]
CAS Number: 37595-74-7
MDL Number: MFCD00000404
Molecular Formula: C8H5F6NO4S2
Molecular Weight: 357.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 102


Product Categories: Synthetic Organic Chemistry, Triflation
Other Product Names / Synonyms: N,N-Bis(trifluoromethylsulfonyl)aniline, Phenyl Triflimide, N-Phenyltrifluoromethanesulfonimide

Formula: C₈H₅F₆NO₄S₂
MW: 357.25 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00000404
CAS Number: 37595-74-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43f54b6c8a301e443f985b1215399bdb3236b33a&vid=6d1e3b7e0a08cd4bc24ef2a46d003d6afdf0fbec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR