Tris(trimethylsilyl)phosphite ≥95.0% (by NMR)
Supplier: TCI America
Synonyms:
Tris(trimethylsilyl)phosphite
P1217-5ML
P1217-25ML
TCP1217-005MLEA
198.23
CAD
TCP1217-005ML
TCP1217-25ML
Tris(trimethylsilyl)phosphite ≥95.0% (by NMR)
Tris(trimethylsilyl) phosphite
CAS Number: 1795-31-9
MDL Number: MFCD00015588
Molecular Formula: C9H27O3PSi3
Molecular Weight: 298.54
Purity/Analysis Method: >95.0% (NMR)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 73
Flash Point (°C): 110
Specific Gravity (20/20): 0.89
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Phosphorous Acid Tris(trimethylsilyl) Ester
Formula:
C₉H₂₇O₃PSi₃ MW: 298.54 g/mol Boiling Pt: 73 °C Density: 0.89 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00015588 CAS Number: 1795-31-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...