1-Pyrenebutyric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1213-1G P1213-5G
TCP1213-001GEA 124.03 CAD
TCP1213-001G TCP1213-005G
1-Pyrenebutyric acid ≥98.0% (by GC, titration analysis)
1-Pyrenebutyric acid

CAS Number: 3443-45-6
MDL Number: MFCD00004141
Molecular Formula: C20H16O2
Molecular Weight: 288.35
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 189

Formula: C₂₀H₁₆O₂
Melting Pt: 189 °C
MDL Number: MFCD00004141
CAS Number: 3443-45-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a147f7eb827fb41aeb61d33e148290c6b0357fb&vid=6a147f7eb827fb41aeb61d33e148290c6b0357fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR