Diethyl (phthalimidomethyl)phosphonate ≥98.0%
Supplier: TCI America
P1193-25G
P1193-5G
TCP1193-25GEA
265.44
CAD
TCP1193-25G
TCP1193-005G
Diethyl (phthalimidomethyl)phosphonate ≥98.0%
Diethyl (phthalimidomethyl)phosphonate
CAS Number: 33512-26-4
MDL Number: MFCD00134313
Molecular Formula: C13H16NO5P
Molecular Weight: 297.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 63
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (Phthalimidomethyl)phosphonic Acid Diethyl Ester
Formula:
C₁₃H₁₆NO₅P Melting Pt: 63 °C |
MDL Number:
MFCD00134313 CAS Number: 33512-26-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...