1H,1H,2H,2H-Perfluoro-1-iodohexane ≥99.0%
Supplier: TCI America
CAS Number: 2043-55-2
MDL Number: MFCD00039409
Molecular Formula: C6H4F9I
Molecular Weight: 373.99
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 126
Melting point (°C): -25
Specific Gravity (20/20): 1.94
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 2-(Nonafluorobutyl)ethyl Iodide, 1H,1H,2H,2H-Perfluorohexyl Iodide
Formula:
C₆H₄F₉I Boiling Pt: 126 °C Melting Pt: –25 °C Density: 1.94 (20 °C) |
MDL Number:
MFCD00039409 CAS Number: 2043-55-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...