Pyromellitic diimide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

P1153-5G P1153-25G
TCP1153-005GEA 102.7 CAD
TCP1153-005G TCP1153-25G
Pyromellitic diimide ≥97.0% (by titrimetric analysis)
Pyromellitic diimide

CAS Number: 2550-73-4
MDL Number: MFCD00005004
Molecular Formula: C10H4N2O4
Molecular Weight: 216.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Benzene-1,2,4,5-tetracarboxylic Diimide

Formula: C₁₀H₄N₂O₄
MDL Number: MFCD00005004
CAS Number: 2550-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87c9099985b37714871a17520353b71eef869cc1&vid=87c9099985b37714871a17520353b71eef869cc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR