Dibenzyl phosphate ≥98.0%
Supplier: TCI America
Synonyms:
Dibenzyl hydrogen phosphate
P1120-25G
P1120-5G
TCP1120-25GEA
584.6
CAD
TCP1120-25G
TCP1120-005G
Dibenzyl phosphate ≥98.0%
Dibenzyl phosphate
CAS Number: 1623-08-1
MDL Number: MFCD00004775
Molecular Formula: C14H15O4P
Molecular Weight: 278.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 81
Storage Temperature: <0°C
Other Product Names / Synonyms: Phosphoric Acid Dibenzyl Ester
Formula:
C₁₄H₁₅O₄P MW: 278.24 g/mol Melting Pt: 81 °C |
MDL Number:
MFCD00004775 CAS Number: 1623-08-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...