N-Phenylurethane ≥99.0%
Supplier: TCI America
P1090-25G
P1090-500G
TCP1090-025GEA
81.05
CAD
TCP1090-025G
TCP1090-500G
N-Phenylurethane ≥99.0%
N-Phenylurethane
CAS Number: 101-99-5
MDL Number: MFCD00026806
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 237
Melting point (°C): 52
Other Product Names / Synonyms: Ethyl Carbanilate, Ethyl N-Phenylcarbamate, N-Phenylcarbamic Acid Ethyl Ester
Formula:
C₉H₁₁NO₂ Boiling Pt: 237 °C Melting Pt: 52 °C |
MDL Number:
MFCD00026806 CAS Number: 101-99-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...