Ethyl (phenylthio)acetate ≥97.0%
Supplier: TCI America
P1085-25G
P1085-5G
TCP1085-25GEA
133.83
CAD
TCP1085-25G
TCP1085-005G
Ethyl (phenylthio)acetate ≥97.0%
Ethyl (phenylthio)acetate
CAS Number: 7605-25-6
MDL Number: MFCD00145107
Molecular Formula: C10H12O2S
Molecular Weight: 196.26
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 160
Specific Gravity (20/20): 1.13
Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: (Phenylthio)acetic Acid Ethyl Ester
Formula:
C₁₀H₁₂O₂S Boiling Pt: 160 °C Density: 1.13 (20 °C) |
MDL Number:
MFCD00145107 CAS Number: 7605-25-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...