2-Pyridylacetic acid hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 2-(Pyridin-2-yl)acetic acid hydrochloride, 2-(Pyridin-2-yl)acetic acid HCl

P1076-5G P1076-25G
TCP1076-005GEA 76 CAD
TCP1076-005G TCP1076-025G
2-Pyridylacetic acid hydrochloride ≥98.0%
2-Pyridylacetic acid hydrochloride

CAS Number: 16179-97-8
MDL Number: MFCD00012812
Molecular Formula: C7H7NO2
Molecular Weight: 173.60
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 130


Other Product Names / Synonyms: 2-Pyridineacetic Acid Hydrochloride

Formula: C₇H₇NO₂
MW: 173.60 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00012812
CAS Number: 16179-97-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d175610d377242f455083b9155ee3872363fae50&vid=d175610d377242f455083b9155ee3872363fae50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR