Methyl-4,4-dimethyl-3-oxovalerate ≥95.0%
Supplier: TCI America
Synonyms:
Pivaloylacetic Acid Methyl Ester, 4,4-Dimethyl-3-oxovaleric Acid Methyl Ester, Methyl 4,4-dimethyl-3-oxopentanoate, Methyl Pivaloylacetate
CAS Number: 55107-14-7
MDL Number: MFCD00008847
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 84
Flash Point (°C): 77
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: 4,4-Dimethyl-3-oxovaleric Acid Methyl Ester, Pivaloylacetic Acid Methyl Ester, Methyl Pivaloylacetate
Formula:
C₈H₁₄O₃ MW: 158.20 g/mol Boiling Pt: 84 °C Density: 0.99 (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00008847 CAS Number: 55107-14-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...