[Hydroxy(tosyloxy)iodo]benzene ≥97.0% (by titrimetric analysis)
Supplier: TCI America
P1015-5G
P1015-25G
TCP1015-005GEA
146.94
CAD
TCP1015-005G
TCP1015-025G
[Hydroxy(tosyloxy)iodo]benzene ≥97.0% (by titrimetric analysis)
[Hydroxy(tosyloxy)iodo]benzene
CAS Number: 27126-76-7
MDL Number: MFCD00011547
Molecular Formula: C13H13IO4S
Molecular Weight: 392.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 138
Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Oxidation
Other Product Names / Synonyms: Phenyliodosohydroxy Tosylate, Koser Reagent
Formula:
C₁₃H₁₃IO₄S Melting Pt: 138 °C |
MDL Number:
MFCD00011547 CAS Number: 27126-76-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...