3,4,9,10-Perylenetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P0972-25G
P0972-500G
P0972-100G
TCP0972-025GEA
49.61
CAD
TCP0972-025G
TCP0972-500G
TCP0972-100G
3,4,9,10-Perylenetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
3,4,9,10-Perylenetetracarboxylic dianhydride
CAS Number: 128-69-8
MDL Number: MFCD00006916
Molecular Formula: C24H8O6
Molecular Weight: 392.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Red
Lambda max.: 470 nm
Product Categories: Materials Chemistry, Organic Transistor Materials, Solar Cell Research Reagents, Conducting Polymer Research Reagents, Monomers, Polyimide Raw Materials, Perylene Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: Pigment Red 224
Formula:
C₂₄H₈O₆ |
MDL Number:
MFCD00006916 CAS Number: 128-69-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...