Bis(2-methoxyethyl)phthalate ≥96.0%

Supplier: TCI America

Synonyms: Bis(2-methoxyethyl) phthalate

P0965-500G P0965-25G
TCP0965-500GEA 140.62 CAD
TCP0965-500G TCP0965-025G
Bis(2-methoxyethyl)phthalate ≥96.0%
Bis(2-methoxyethyl) phthalate

CAS Number: 117-82-8
MDL Number: MFCD00042842
Molecular Formula: C14H18O6
Molecular Weight: 282.29
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 340
Flash Point (°C): 185
Freezing point (°C): -45
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: Phthalic Acid Bis(2-methoxyethyl) Ester

Formula: C₁₄H₁₈O₆
MW: 282.29 g/mol
Boiling Pt: 340 °C
Density: 1.17 (20 °C)
Flash Pt: 185 °C
MDL Number: MFCD00042842
CAS Number: 117-82-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65ada4d9985fbe078ab75ddb2d29b1b6cf246ac7&vid=65ada4d9985fbe078ab75ddb2d29b1b6cf246ac7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR