N-Phthaloylglycine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0963-500G P0963-25G
TCP0963-500GEA 285.98 CAD
TCP0963-500G TCP0963-025G
N-Phthaloylglycine ≥98.0% (by HPLC, titration analysis)
N-Phthaloylglycine

CAS Number: 4702-13-0
MDL Number: MFCD00005900
Molecular Formula: C10H7NO4
Molecular Weight: 205.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 196


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Phthalimidoacetic Acid

Formula: C₁₀H₇NO₄
Melting Pt: 196 °C
MDL Number: MFCD00005900
CAS Number: 4702-13-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR