trans-4-Pentylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

P0944-5G P0944-25G
TCP0944-005GEA 71.1 CAD
TCP0944-005G TCP0944-025G
trans-4-Pentylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)
trans-4-Pentylcyclohexanecarboxylic acid

CAS Number: 38289-29-1
MDL Number: MFCD00009964
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 55
Flash Point (°C): 110


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: trans-4-Amylcyclohexanecarboxylic Acid

Formula: C₁₁H₂₀O₂
Melting Pt: 55 °C
Flash Pt: 110 °C
MDL Number: MFCD00009964
CAS Number: 38289-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98f13e983c0d8a25ae0a5802e7380a504e65505d&vid=98f13e983c0d8a25ae0a5802e7380a504e65505d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR