Cobalt(II) Phthalocyanine ≥93.0%
Supplier: TCI America
Synonyms:
Cobalt(II) phthalocyanine, Phthalocyanine Cobalt(II), 29H,31H-Phthalocyanine, cobalt complex
CAS Number: 3317-67-7
MDL Number: MFCD00010718
Molecular Formula: C32H16CoN8
Molecular Weight: 571.47
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Color: Deep Red
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Molecular Conductors, Magnetic Metal Complexes (Magnetic Materials), Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Phthalocyanine Cobalt(II)
Formula:
C₃₂H₁₆CoN₈ MW: 571.47 g/mol |
MDL Number:
MFCD00010718 CAS Number: 3317-67-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...