Polyethylene glycol mono-4-octylphenyl ether, M.W. n=∼10

Supplier: TCI America

Synonyms: Triton X-100, Polyethylene glycol mono(4-tert-octylphenyl) ether

P0873-25G P0873-500G
TCP0873-025GEA 39.5 CAD
TCP0873-025G TCP0873-500G
Polyethylene glycol mono-4-octylphenyl ether, M.W. n=∼10
Polyethylene glycol mono-4-octylphenyl ether

CAS Number: 9002-93-1
MDL Number: MFCD00132505
Form: Clear Liquid
Color: Colorless
Melting point (°C): 6
Flash Point (°C): 286
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Nonionic Surfactants

Formula: C8H17C6H4(OCH2CH2)nOH
Melting Pt: 6 °C
Density: 1.07 (20 °C)
Flash Pt: 286 °C
MDL Number: MFCD00132505
CAS Number: 9002-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce22e6aa971860788d1735b7365f04a348dc6f74&vid=32cecd182803afa2348ef9fe0b920ffa91ac0123&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR