Ethyl phenylpropiolate ≥97.0%

Supplier: TCI America

P0814-5G P0814-25G
TCP0814-5GEA 69.36 CAD
TCP0814-5G TCP0814-25G
Ethyl phenylpropiolate ≥97.0%
Ethyl phenylpropiolate

CAS Number: 2216-94-6
MDL Number: MFCD00009185
Molecular Formula: C11H10O2
Molecular Weight: 174.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 110
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Phenylpropiolic Acid Ethyl Ester, Ethyl Phenylpropargylate, Phenylpropargylic Acid Ethyl Ester

Formula: C₁₁H₁₀O₂
Boiling Pt: 103 °C
Density: 1.06 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009185
CAS Number: 2216-94-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f39d5287942192ddce1030631f1c597eb88225c3&vid=f39d5287942192ddce1030631f1c597eb88225c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR