Polyvinyl alcohol n ∼2,000, (degree of saponification ca. 80mol%)

Supplier: TCI America

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

P0804-500G P0804-25G
TCP0804-500GEA 127.84 CAD
TCP0804-500G TCP0804-025G
Polyvinyl alcohol n ∼2,000, (degree of saponification ca. 80mol%)
Polyvinyl alcohol

Form: Crystals
Color: White
Flash Point (°C): 77


Product Categories: Materials Chemistry, Polymer Research Reagents

Formula: (CH₂CHOH)n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Solubility in Water almost transparency
degree's of saponification 78.0 to 82.0 mol%
Suitability for Absorptiometry to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR