Diphenyl phosphate ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

P0801-1G P0801-25G
TCP0801-001GEA 41.08 CAD
TCP0801-001G TCP0801-025G
Diphenyl phosphate ≥99.0% (by HPLC, titration analysis)
Diphenyl phosphate

CAS Number: 838-85-7
MDL Number: MFCD00003033
Molecular Formula: C12H11O4P
Molecular Weight: 250.19
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 69
Flash Point (°C): 302


Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Phosphoric Acid Diphenyl Ester

Formula: C₁₂H₁₁O₄P
Melting Pt: 69 °C
Flash Pt: 302 °C
MDL Number: MFCD00003033
CAS Number: 838-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e0d509d4fc95c495eed741b507405ca01cbd340&vid=1e0d509d4fc95c495eed741b507405ca01cbd340&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR