1-Phenacylpyridinium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0797-5G P0797-25G
TCP0797-005GEA 42.66 CAD
TCP0797-005G TCP0797-025G
1-Phenacylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Phenacylpyridinium bromide

CAS Number: 16883-69-5
MDL Number: MFCD00031995
Molecular Formula: C13H12BrNO
Molecular Weight: 278.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 198


Other Product Names / Synonyms: 1-Acetophenonepyridinium Bromide

Formula: C₁₃H₁₂BrNO
Melting Pt: 198 °C
MDL Number: MFCD00031995
CAS Number: 16883-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ee3525332f560bc0cf0500c91b969336b8ff1fb&vid=6ee3525332f560bc0cf0500c91b969336b8ff1fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR