Iron(II) phthalocyanine ≥95.0% (by titrimetric analysis)

Supplier: TCI America

P0774-25G P0774-5G
TCP0774-025GEA 183.28 CAD
TCP0774-025G TCP0774-5G
Iron(II) phthalocyanine ≥95.0% (by titrimetric analysis)
Iron(II) phthalocyanine

CAS Number: 132-16-1
MDL Number: MFCD00015953
Molecular Formula: C32H16FeN8
Molecular Weight: 568.38
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Molecular Conductors, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Phthalocyanine Iron(II)

Formula: C₃₂H₁₆FeN₈
MDL Number: MFCD00015953
CAS Number: 132-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3332f292f5ab155f2e11d529b2e6b6bbe46c004c&vid=3332f292f5ab155f2e11d529b2e6b6bbe46c004c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR